Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019336.1 | NP_062209.1 | 4 | 463 | ||
XM_006249974.2 | XP_006250036.1 | 4 | 647 | |||
GenBank mRNA | BC098681.1 | - | 4 | 463 | 7385088 | |
FQ234088.1 | - | 4 | 361 | |||
U77698.1 | - | 1 | 138 |