Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242933.1 | NP_001229862.1 | 13 | 1550 | ||
NM_003099.4 | NP_003090.2 | 13 | 1550 | |||
NM_148955.3 | NP_683758.1 | 11 | 1355 | |||
XM_005254611.2 | XP_005254668.1 | 13 | 1526 | |||
GenBank mRNA | AB209013.1 | - | 13 | 1357 | ||
AF065483.1 | - | 13 | 1420 | |||
AF065484.1 | - | 11 | 1225 | |||
AK128179.1 | - | 13 | 1442 | |||
AK222561.1 | - | 13 | 1434 | |||
AK222793.1 | - | 13 | 1423 | |||
AK291752.1 | - | 13 | 1434 | |||
BC000357.2 | - | 13 | 1430 | 2964409 | ||
BT006983.1 | - | 13 | 1420 | |||
U53225.1 | - | 13 | 1465 |