Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183328.2 | NP_899157.1 | 4 | 300 | ||
XM_008762822.1 | XP_008761044.1 | 5 | 546 | |||
XM_008762823.1 | XP_008761045.1 | 5 | 482 | |||
XM_008762824.1 | XP_008761046.1 | 5 | 426 | |||
XM_008762825.1 | XP_008761047.1 | 5 | 414 | |||
XM_008762826.1 | XP_008761048.1 | 5 | 365 | |||
XM_008762827.1 | XP_008761049.1 | 5 | 546 | |||
XM_008762828.1 | XP_008761050.1 | 5 | 421 | |||
XM_008762829.1 | XP_008761051.1 | 4 | 345 | |||
GenBank mRNA | M94557.1 | - | 4 | 354 |