Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMHR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164690.1 | NP_001158162.1 | 8 | 1099 | ||
NM_001164691.1 | NP_001158163.1 | 8 | 1099 | |||
NM_020547.2 | NP_065434.1 | 8 | 1099 | |||
XM_011538173.1 | XP_011536475.1 | 9 | 1159 | |||
XM_011538174.1 | XP_011536476.1 | 9 | 1156 | |||
XM_011538176.1 | XP_011536478.1 | 8 | 1102 | |||
XM_011538178.1 | XP_011536480.1 | 9 | 940 | |||
XM_011538179.1 | XP_011536481.1 | 9 | 1159 | |||
XM_011538180.1 | XP_011536482.1 | 9 | 1073 | |||
XM_011538181.1 | XP_011536483.1 | 9 | 1070 | |||
XM_011538183.2 | XP_011536485.1 | 9 | 1159 | |||
XM_011538184.2 | XP_011536486.1 | 9 | 1159 | |||
XM_011538186.2 | XP_011536488.1 | 4 | 296 | |||
XM_011538187.2 | XP_011536489.1 | 7 | 987 | |||
XM_017019179.1 | XP_016874668.1 | 9 | 1159 | |||
GenBank mRNA | AF172932.1 | - | 8 | 1082 | ||
AK313593.1 | - | 8 | 1088 | |||
AY714878.1 | - | 8 | 1019 | |||
BC126316.1 | - | 8 | 1074 | 8992032 | ||
BC136356.1 | - | 8 | 1074 | 9020343 | ||
CR627318.1 | - | 4 | 402 |