Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sstr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019348.1 | NP_062221.1 | 2 | 1421 | ||
GenBank mRNA | M93273.1 | - | 2 | 1421 | ||
M96817.1 | - | 2 | 1421 |
Set Membership: |
G-protein coupled receptor |