Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019363.3 | NP_062236.2 | 30 | 3445 | ||
GenBank mRNA | AF110477.1 | - | 30 | 3462 | ||
AF110478.1 | - | 30 | 3446 |