Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rassf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019365.3 | NP_062238.1 | 1, 2 | 568 | ||
GenBank mRNA | AF002251.1 | - | 1, 2 | 568 |