Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019367.2 | NP_062240.1 | 5 | 621 | ||
XM_006256017.1 | XP_006256079.1 | 5 | 936 | |||
XM_006256018.2 | XP_006256080.1 | 5 | 1758 | |||
XM_006256019.2 | XP_006256081.1 | 5 | 1491 | |||
XM_006256020.2 | XP_006256082.1 | 5 | 1346 | |||
XM_006256022.2 | XP_006256084.1 | 5 | 1305 | |||
XM_006256023.2 | XP_006256085.1 | 5 | 779 | |||
XM_008772762.1 | XP_008770984.1 | 5 | 2245 | |||
GenBank mRNA | AF061971.1 | - | 5 | 553 | ||
BC062023.1 | - | 5 | 583 | 5623515 | ||
FQ234132.1 | - | 5 | 563 |