Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019373.2 | NP_062246.1 | 3 | 415 | ||
GenBank mRNA | AF207821.1 | - | 3 | 392 | ||
BC089807.1 | - | 3 | 364 | 7323885 | ||
FQ210093.1 | - | 3 | 414 | |||
FQ219449.1 | - | 3 | 415 |