Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005632.2 | NP_005623.1 | 14 | 4194 | ||
XM_011522620.2 | XP_011520922.1 | 15 | 7517 | |||
XM_011522621.2 | XP_011520923.1 | 15 | 8445 | |||
XM_011522622.1 | XP_011520924.1 | 13 | 7149 | |||
XM_011522623.2 | XP_011520925.1 | 16 | 5220 | |||
XM_011522624.2 | XP_011520926.1 | 15 | 4558 | |||
XM_011522625.2 | XP_011520927.1 | 14 | 4444 | |||
XM_011522626.2 | XP_011520928.1 | 15 | 7985 | |||
XM_011522627.2 | XP_011520929.1 | 15 | 7986 | |||
XM_011522628.2 | XP_011520930.1 | 15 | 5078 | |||
XM_011522629.2 | XP_011520931.1 | 15 | 4941 | |||
XM_011522630.2 | XP_011520932.1 | 14 | 4279 | |||
XM_011522631.2 | XP_011520933.1 | 13 | 4165 | |||
XM_011522632.2 | XP_011520934.1 | 13 | 4596 | |||
XM_017023596.1 | XP_016879085.1 | 16 | 5357 | |||
GenBank mRNA | BC001869.1 | - | 2 | 920 | ||
BC021854.1 | - | 10 | 2261 | |||
BC040169.1 | - | 1 | 390 | |||
BC112918.1 | - | 6 | 2419 | |||
U69557.1 | - | 2 | 880 |