Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SON | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291411.1 | NP_001278340.1 | 3 | 3365 | ||
NM_032195.2 | NP_115571.2 | 3 | 3365 | |||
NM_138927.2 | NP_620305.2 | 3 | 3365 | |||
NR_103797.1 | 3 | 3365 | ||||
GenBank mRNA | AB028942.2 | - | 3 | 3334 | ||
AF380179.1 | - | 4 | 2382 | |||
AF380180.1 | - | 3 | 3339 | |||
AF380181.1 | - | 3 | 3339 | |||
AF380183.1 | - | 3 | 3339 | |||
AF380184.1 | - | 3 | 3339 | |||
AK074269.1 | - | 1 | 2327 | |||
AK128071.1 | - | 1 | 2312 | |||
AK307612.1 | - | 3 | 3365 | |||
AY026895.1 | - | 4 | 3239 | |||
BC002422.2 | - | 1 | 1829 | |||
X63751.1 | - | 1 | 1306 | |||
X63753.1 | - | 2 | 700 |