Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Park2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020093.1 | NP_064478.1 | 8, 9 | 1000 | ||
GenBank mRNA | AB039878.1 | - | 8, 9 | 1000 | ||
AF168004.1 | - | 8, 9 | 1020 | |||
AF210434.1 | - | 8, 9 | 992 | |||
AF257234.1 | - | 7, 8 | 930 | |||
AF343574.1 | - | 8, 9 | 689 | |||
AF381277.1 | - | 8, 9 | 1002 | |||
AF381278.1 | - | 7, 8 | 930 | |||
AF381279.1 | - | 6, 7 | 808 | |||
AF381280.1 | - | 8, 9 | 1086 | |||
AF381281.1 | - | 6, 7 | 846 | |||
AF381285.1 | - | 1, 2 | 66 | |||
GU322017.1 | - | 9, 10 | 1099 | |||
GU322019.1 | - | 9, 10 | 1030 | |||
GU322363.1 | - | 7, 8 | 859 | |||
GU322364.1 | - | 8, 9 | 943 | |||
GU345835.1 | - | 9, 10 | 1177 | |||
GU345836.1 | - | 9, 10 | 978 | |||
KC774169.1 | - | 7, 8 | 786 | |||
KC774170.1 | - | 7, 8 | 786 | |||
KC774172.1 | - | 8, 9 | 940 | |||
KC774174.1 | - | 3, 4 | 223 | |||
KC774176.1 | - | 5, 6 | 587 |