Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033961.1 | NP_001029133.1 | 2 | 371 | ||
NM_020094.2 | NP_064479.2 | 4 | 521 | |||
NM_020095.2 | NP_064480.2 | 3 | 467 | |||
XM_008760448.1 | XP_008758670.1 | 2 | 2079 | |||
XM_008760449.1 | XP_008758671.1 | 4 | 630 | |||
XM_008760450.1 | XP_008758672.1 | 2 | 271 | |||
GenBank mRNA | AF269283.1 | - | 4 | 299 | ||
AF269284.1 | - | 3 | 245 | |||
AF269285.1 | - | 2 | 149 | |||
BC085905.1 | - | 2 | 371 | 7193750 | ||
GU937871.1 | - | 2 | 181 |
Gene Symbol: LOC100911951 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006231581.2 | XP_006231643.1 | 2 | 267 | ||
XM_008760479.1 | XP_008758701.1 | 4 | 578 | |||
XM_008760480.1 | XP_008758702.1 | 3 | 523 | |||
XM_008760481.1 | XP_008758703.1 | 2 | 585 | |||
XM_008760482.1 | XP_008758704.1 | 2 | 426 | |||
XM_008760483.1 | XP_008758705.1 | 4 | 630 |