Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000036.2 | NP_000027.2 | 4 | 509 | ||
NM_001172626.1 | NP_001166097.1 | 3 | 497 | |||
GenBank mRNA | AK097077.1 | - | 3 | 497 | ||
AK291349.1 | - | 4 | 413 | |||
AK314252.1 | - | 4 | 419 | |||
BC056678.1 | - | 4 | 438 | |||
DA826076.1 | - | 4 | 478 | |||
M60092.1 | - | 4 | 446 |