Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SORL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003105.5 | NP_003096.1 | 40 | 5466 | ||
XM_011542963.2 | XP_011541265.1 | 39 | 5227 | |||
XM_011542965.2 | XP_011541267.1 | 34 | 4375 | |||
XM_011542967.2 | XP_011541269.1 | 20 | 2407 | |||
XM_017018169.1 | XP_016873658.1 | 40 | 5173 | |||
XM_017018170.1 | XP_016873659.1 | 38 | 4849 | |||
XM_017018172.1 | XP_016873661.1 | 26 | 3338 | |||
GenBank mRNA | AK294404.1 | - | 11 | 1604 | ||
AK295090.1 | - | 17 | 2256 | |||
AK295878.1 | - | 20 | 2407 | |||
AK302589.1 | - | 17 | 2078 | |||
AK316340.1 | - | 11 | 1186 | |||
BC137171.1 | - | 40 | 5389 | 9021168 | ||
BQ888333.1 | - | 7 | 688 | |||
Y08110.1 | - | 40 | 5417 |