Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005633.3 | NP_005624.2 | 6 | 789 | ||
XM_005264515.4 | XP_005264572.1 | 6 | 1018 | |||
XM_011533062.2 | XP_011531364.1 | 6 | 957 | |||
XM_011533064.2 | XP_011531366.1 | 6 | 697 | |||
GenBank mRNA | AK290228.1 | - | 7 | 835 | ||
AK301960.1 | - | 6 | 848 | |||
AL833457.1 | - | 1 | 628 | |||
L13857.1 | - | 6 | 748 |