Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zranb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031616.1 | NP_113804.1 | 5 | 351 | ||
GenBank mRNA | AF013965.1 | - | 5 | 591 | ||
AF013967.1 | - | 5 | 351 | |||
BC087012.1 | - | 5 | 337 | 7189430 | ||
FQ211774.1 | - | 5 | 377 | |||
FQ211917.1 | - | 5 | 377 |