Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031619.2 | NP_113807.2 | 3 | 409 | ||
GenBank mRNA | D50568.1 | - | 3 | 409 |