Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209277.1 | - | 4 | 464 | ||
AI419975.1 | - | 4 | 472 | |||
BC117261.1 | - | 4 | 400 | 40125812 | ||
BC143366.1 | - | 4 | 400 | 9051871 | ||
BC143367.1 | - | 4 | 400 | 9051872 | ||
L13858.1 | - | 4 | 374 |