Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fut2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031635.1 | NP_113823.1 | 3 | 1190 | ||
XM_006229130.2 | XP_006229192.1 | 2 | 1086 | |||
XM_006229135.2 | XP_006229197.1 | 3 | 1178 | |||
XM_006229136.2 | XP_006229198.1 | 2 | 1089 | |||
XM_006229137.2 | XP_006229199.1 | 2 | 3938 | |||
XM_006229138.2 | XP_006229200.1 | 2 | 2344 | |||
XM_006229139.2 | XP_006229201.1 | 2 | 4199 | |||
XM_006229140.2 | XP_006229202.1 | 2 | 1065 | |||
XM_006229141.2 | XP_006229203.1 | 2 | 2334 | |||
XM_008759412.1 | XP_008757634.1 | 2 | 1197 | |||
GenBank mRNA | AB006138.1 | - | 3 | 1190 | ||
AF042743.1 | - | 1 | 896 | |||
AF264005.1 | - | 1 | 977 | |||
L26010.1 | - | 1 | 316 |