Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fut2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031635.1 | NP_113823.1 | 3 | 339 | ||
XM_006229130.2 | XP_006229192.1 | 2 | 235 | |||
XM_006229135.2 | XP_006229197.1 | 3 | 327 | |||
XM_006229136.2 | XP_006229198.1 | 2 | 238 | |||
XM_006229137.2 | XP_006229199.1 | 2 | 3087 | |||
XM_006229138.2 | XP_006229200.1 | 2 | 1493 | |||
XM_006229139.2 | XP_006229201.1 | 2 | 3348 | |||
XM_006229140.2 | XP_006229202.1 | 2 | 214 | |||
XM_006229141.2 | XP_006229203.1 | 2 | 1483 | |||
XM_008759412.1 | XP_008757634.1 | 2 | 346 | |||
GenBank mRNA | AB006138.1 | - | 3 | 339 | ||
AF042743.1 | - | 1 | 45 | |||
AF264005.1 | - | 1 | 126 |