Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031639.1 | NP_113827.1 | 21 | 2816 | ||
XM_006257080.2 | XP_006257142.1 | 21 | 3143 | |||
XM_006257081.2 | XP_006257143.1 | 20 | 3088 | |||
XM_006257082.2 | XP_006257144.1 | 20 | 3051 | |||
XM_006257083.2 | XP_006257145.1 | 19 | 2995 | |||
XM_006257084.2 | XP_006257146.1 | 14 | 1543 | |||
XM_006257085.2 | XP_006257147.1 | 12 | 1934 | |||
XM_008773267.1 | XP_008771489.1 | 20 | 3103 | |||
XM_008773268.1 | XP_008771490.1 | 14 | 1452 | |||
GenBank mRNA | U50147.1 | - | 21 | 2816 |