Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCR11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006830.3 | NP_006821.1 | 3 | 541 | ||
GenBank mRNA | BC000462.2 | - | 3 | 489 | 2822699 | |
BC003594.1 | - | 3 | 489 | 2822699 |