Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crhr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247542.2 | XP_006247604.1 | 9 | 2182 | ||
GenBank mRNA | EU012436.1 | - | 10 | 908 | ||
EU012437.1 | - | 9 | 757 | |||
EU012438.1 | - | 10 | 976 | |||
L24096.1 | - | 10 | 989 | |||
L25438.1 | - | 10 | 911 | |||
U53500.1 | - | 7 | 541 |
Set Membership: |
G-protein coupled receptor |