Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ramp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031646.1 | NP_113834.1 | 3, 4 | 300 | ||
GenBank mRNA | AB028934.1 | - | 1, 2 | 14 | ||
AB030943.1 | - | 3, 4 | 309 | |||
AB042888.1 | - | 3, 4 | 300 | |||
AF162778.1 | - | 1, 2 | 54 | |||
AF181551.1 | - | 3, 4 | 303 | |||
EF595744.1 | - | 3, 4 | 224 | |||
FQ226087.1 | - | 3, 4 | 325 |