Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ltb4r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021656.1 | NP_067688.1 | 1 | 433 | ||
XM_006252011.2 | XP_006252073.1 | 2 | 2107 | |||
XM_006252012.1 | XP_006252074.1 | 2 | 469 |
Set Membership: |
G-protein coupled receptor |