Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smad5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021692.1 | NP_067724.1 | 5 | 1150 | ||
XM_006253586.2 | XP_006253648.1 | 8 | 1615 | |||
XM_006253587.2 | XP_006253649.1 | 7 | 1533 | |||
XM_006253588.2 | XP_006253650.1 | 8 | 1857 | |||
XM_008771446.1 | XP_008769668.1 | 7 | 3193 | |||
XM_008771447.1 | XP_008769669.1 | 8 | 2375 | |||
XM_008771448.1 | XP_008769670.1 | 8 | 2550 | |||
GenBank mRNA | AB010955.1 | - | 5 | 1150 | ||
BC161849.1 | - | 6 | 1337 | 9099068 |