Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021697.1 | NP_067729.1 | 3 | 1199 | ||
GenBank mRNA | FQ211837.1 | - | 1210 | |||
X98564.1 | - | 3 | 1199 |