Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021702.1 | NP_067734.1 | 4 | 381 | ||
XM_006240485.2 | XP_006240547.1 | 5 | 417 | |||
XM_006240486.2 | XP_006240548.1 | 5 | 414 | |||
XM_006240487.2 | XP_006240549.1 | 5 | 417 | |||
XM_006240488.2 | XP_006240550.1 | 4 | 333 | |||
XM_006240490.1 | XP_006240552.1 | 5 | 443 | |||
XM_006240493.2 | XP_006240555.1 | 4 | 336 | |||
GenBank mRNA | Y12319.1 | - | 4 | 381 |