Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021703.1 | NP_067735.1 | 6 | 1673 | ||
GenBank mRNA | AJ002474.1 | - | 6 | 1673 |