Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270688.1 | NP_001257617.1 | 5 | 278 | ||
NM_001270689.1 | NP_001257618.1 | 5 | 306 | |||
NM_021909.2 | NP_068709.2 | 5 | 382 | |||
GenBank mRNA | BC060571.1 | - | 5 | 270 | 6891214 | |
DV726876.1 | - | 1 | 48 | |||
FM122563.1 | - | 5 | 256 | |||
FQ220058.1 | - | 5 | 306 |