Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col5a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021760.1 | NP_068528.1 | 63 | 4594 | ||
XM_008765979.1 | XP_008764201.1 | 63 | 4659 | |||
GenBank mRNA | AF272661.1 | - | 63 | 4594 |