Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324113.1 | NP_001311042.1 | 11 | 1751 | ||
NM_001324114.1 | NP_001311043.1 | 10 | 1639 | |||
NM_001324115.1 | NP_001311044.1 | 11 | 1978 | |||
NM_001324116.1 | NP_001311045.1 | 11 | 1917 | |||
NM_001324117.1 | NP_001311046.1 | 11 | 1914 | |||
NM_003115.4 | NP_003106.3 | 10 | 1800 | |||
XM_011509913.2 | XP_011508215.1 | 12 | 1750 | |||
XM_011509914.2 | XP_011508216.1 | 12 | 1802 | |||
GenBank mRNA | AB011004.1 | - | 9 | 1471 | ||
AK312685.1 | - | 10 | 1726 | |||
BC009377.2 | - | 10 | 1773 | 4121439 | ||
S73498.1 | - | 10 | 1703 |