Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021850.2 | NP_068622.1 | 3 | 275 | ||
XM_006252016.2 | XP_006252078.1 | 3 | 465 | |||
XM_008770692.1 | XP_008768914.1 | 2 | 746 | |||
XM_008770693.1 | XP_008768915.1 | 2 | 822 | |||
XM_008770694.1 | XP_008768916.1 | 3 | 885 | |||
GenBank mRNA | AF096291.1 | - | 1 | 77 | ||
AY185098.1 | - | 3 | 246 | |||
AY185100.1 | - | 1 | 585 | |||
BC074021.1 | - | 3 | 275 | 7105621 | ||
FQ212018.1 | - | 3 | 432 |