Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174044.1 | NP_001167515.1 | 5 | 1678 | ||
NM_001174045.1 | NP_001167516.1 | 6 | 1673 | |||
NM_001174046.1 | NP_001167517.1 | 5 | 1588 | |||
NM_003117.4 | NP_003108.2 | 5 | 1499 | |||
NM_153189.2 | NP_694859.1 | 5 | 1499 | |||
GenBank mRNA | AK292229.1 | - | 5 | 1588 | ||
AY920278.1 | - | 5 | 1674 | |||
AY920279.1 | - | 5 | 1495 | |||
AY920280.1 | - | 6 | 1648 | |||
AY920281.1 | - | 5 | 1563 | |||
BC026163.1 | - | 5 | 1422 | 4838230 | ||
EU794686.1 | - | 5 | 1462 | |||
L13781.1 | - | 3 | 1161 | |||
S67798.1 | - | 5 | 1433 | |||
X84347.1 | - | 4 | 1356 |