Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080547.1 | NP_001074016.1 | 2 | 303 | ||
NM_003120.2 | NP_003111.2 | 2 | 300 | |||
XM_011520307.1 | XP_011518609.1 | 4 | 1745 | |||
XM_017018173.1 | XP_016873662.1 | 4 | 1748 | |||
GenBank mRNA | AK310065.1 | - | 2 | 211 | ||
BI909430.1 | - | 2 | 104 | |||
BP271276.1 | - | 2 | 300 | |||
X52056.1 | - | 2 | 270 |