Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022209.3 | NP_071545.2 | 9 | 1320 | ||
GenBank mRNA | AY251277.1 | - | 9 | 1272 | ||
BC078834.1 | - | 12 | 1529 | 7114294 | ||
D38260.1 | - | 9 | 1115 | |||
FQ212581.1 | - | 7 | 840 |