Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243998.1 | NP_001230927.1 | 4 | 192 | ||
NM_001243999.1 | NP_001230928.1 | 5 | 407 | |||
NM_001244000.1 | NP_001230929.1 | 4, 5 | 372 | |||
NM_003121.4 | NP_003112.2 | 5 | 407 | |||
GenBank mRNA | AK225225.1 | - | 5 | 405 | ||
AK225720.1 | - | 5 | 407 | |||
AK292644.1 | - | 5 | 409 | |||
AK300639.1 | - | 4 | 190 | |||
BC007921.2 | - | 5 | 386 | 4309499 | ||
DA957263.1 | - | 4, 5 | 369 | |||
X66079.1 | - | 5 | 387 | |||
X96998.1 | - | 5 | 387 |