Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000480.2 | NP_000471.1 | 8 | 1584 | ||
NM_001025389.1 | NP_001020560.1 | 8 | 1547 | |||
NM_001025390.1 | NP_001020561.1 | 8 | 1686 | |||
NM_001172430.1 | NP_001165901.1 | 8 | 1511 | |||
NM_001172431.1 | NP_001165902.1 | 7 | 1207 | |||
GenBank mRNA | AK289998.1 | - | 8 | 1318 | ||
AK294444.1 | - | 6 | 909 | |||
AK295046.1 | - | 7 | 1205 | |||
AK299113.1 | - | 8 | 1259 | |||
AK301507.1 | - | 8 | 1434 | |||
AK302970.1 | - | 8 | 1502 | |||
AK313039.1 | - | 8 | 1232 | |||
BC126118.1 | - | 8 | 1579 | 8991834 | ||
D12775.1 | - | 8 | 1403 | |||
M84721.1 | - | 8 | 1346 | |||
M84722.1 | - | 8 | 1470 |