Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPINT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032367.1 | NP_001027539.1 | 2 | 621 | ||
NM_003710.3 | NP_003701.1 | 3 | 715 | |||
NM_181642.2 | NP_857593.1 | 3 | 715 | |||
XM_006720657.1 | XP_006720720.1 | 3 | 678 | |||
XM_011521957.1 | XP_011520259.1 | 3 | 745 | |||
XM_011521958.1 | XP_011520260.1 | 3 | 745 | |||
GenBank mRNA | AB000095.1 | - | 3 | 686 | ||
AK300195.1 | - | 4 | 616 | |||
AK303899.1 | - | 3 | 461 | |||
AK314825.1 | - | 3 | 746 | |||
AY296715.1 | - | 3 | 598 | |||
AY358969.1 | - | 3 | 698 | |||
BC004140.1 | - | 2 | 563 | 2959383 | ||
BC018702.1 | - | 3 | 707 | 3140292 | ||
BT007425.1 | - | 2 | 511 |