Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc33a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022252.2 | NP_071588.2 | 6 | 1759 | ||
XM_008761058.1 | XP_008759280.1 | 6 | 2183 | |||
GenBank mRNA | BC078832.1 | - | 6 | 1759 | 7132173 |