Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022264.1 | NP_071600.1 | 2 | 249 | ||
XM_006250909.2 | XP_006250971.1 | 2 | 283 | |||
GenBank mRNA | AF228307.1 | - | 2 | 226 | ||
AF228308.1 | - | 2 | 223 | |||
AF228309.1 | - | 2 | 226 | |||
AF228311.1 | - | 2 | 226 | |||
AF296694.1 | - | 2 | 233 | |||
D12524.1 | - | 2 | 249 | |||
EU247827.1 | - | 2 | 233 | |||
EU247828.1 | - | 2 | 233 |