Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199828.1 | NP_001186757.1 | 2 | 465 | ||
NM_005987.3 | NP_005978.2 | 2 | 451 |