Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xylt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006230151.2 | XP_006230213.2 | 12 | 2359 | ||
GenBank mRNA | AJ295748.1 | - | 9 | 1865 |