Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uba52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031687.3 | NP_113875.1 | 4 | 319 | ||
XM_006252934.2 | XP_006252996.1 | 4 | 406 | |||
XM_006252935.1 | XP_006252997.1 | 4 | 524 | |||
XM_008771145.1 | XP_008769367.1 | 4 | 402 | |||
XM_008771146.1 | XP_008769368.1 | 4 | 346 | |||
GenBank mRNA | BC061544.1 | - | 4 | 315 | 6922034 | |
BP492585.1 | - | 3 | 205 | |||
FQ209944.1 | - | 4 | 311 | |||
FQ210541.1 | - | 4 | 308 | |||
FQ210904.1 | - | 4 | 309 | |||
FQ212001.1 | - | 4 | 313 | |||
FQ212205.1 | - | 4 | 311 | |||
FQ212291.1 | - | 4 | 309 | |||
FQ212810.1 | - | 4 | 312 | |||
FQ217153.1 | - | 4 | 320 | |||
FQ217440.1 | - | 4 | 304 | |||
FQ219376.1 | - | 4 | 310 | |||
FQ219803.1 | - | 4 | 311 | |||
FQ221045.1 | - | 4 | 308 | |||
FQ221118.1 | - | 4 | 309 | |||
FQ221238.1 | - | 4 | 529 | |||
FQ221621.1 | - | 4 | 305 | |||
FQ221631.1 | - | 4 | 311 | |||
FQ221755.1 | - | 4 | 309 | |||
FQ221772.1 | - | 4 | 309 | |||
FQ221777.1 | - | 4 | 309 | |||
FQ222045.1 | - | 4 | 309 | |||
FQ222098.1 | - | 4 | 305 | |||
FQ222371.1 | - | 4 | 308 | |||
FQ222483.1 | - | 4 | 308 | |||
FQ222507.1 | - | 4 | 303 | |||
FQ222550.1 | - | 4 | 305 | |||
FQ222700.1 | - | 4 | 308 | |||
FQ223258.1 | - | 4 | 311 | |||
FQ223423.1 | - | 4 | 305 | |||
FQ224647.1 | - | 3 | 299 | |||
FQ224702.1 | - | 4 | 309 | |||
FQ228362.1 | - | 4 | 305 | |||
FQ228566.1 | - | 4 | 309 | |||
U25064.1 | - | 1 | 42 | |||
X82636.1 | - | 3 | 281 |