Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022383.2 | NP_071778.2 | 5 | 487 | ||
XM_006238830.2 | XP_006238892.1 | 5 | 587 | |||
XM_006238831.2 | XP_006238893.1 | 5 | 584 | |||
XM_006238832.2 | XP_006238894.1 | 5 | 565 | |||
GenBank mRNA | BC089801.1 | - | 5 | 487 | 7381845 | |
L11930.1 | - | 4 | 389 |