Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RPLP0 SIRNA Assays ›
Gene Symbol:
Rplp0
Gene Name:
ribosomal protein, large, P0
Gene Aliases:
Arbp
Chromosome Location:
Chr.12: 46791528 - 46794797 on Build Rnor_6.0
UniGene ID:
Rn.1079
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s133912
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Rplp0
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022402.2
NP_071797.1
5
632
GenBank mRNA
BC062028.1
-
4
601
5623881
DN931898.1
-
4
650
FQ209537.1
-
5
632
FQ209603.1
-
5
632
FQ209620.1
-
4
617
FQ209757.1
-
5
633
FQ210253.1
-
5
632
FQ210412.1
-
5
632
FQ211152.1
-
5
632
FQ212554.1
-
5
632
FQ212743.1
-
5
632
FQ212875.1
-
5
632
FQ212927.1
-
5
633
FQ213085.1
-
5
632
FQ213427.1
-
5
632
FQ213510.1
-
5
632
FQ213712.1
-
5
632
FQ213847.1
-
5
633
FQ214087.1
-
5
632
FQ214515.1
-
5
632
FQ214553.1
-
5
633
FQ214850.1
-
5
632
FQ214997.1
-
5
632
FQ215007.1
-
5
632
FQ215548.1
-
5
634
FQ215662.1
-
5
632
FQ215682.1
-
5
632
FQ215870.1
-
5
632
FQ216120.1
-
5
633
FQ216296.1
-
5
632
FQ216389.1
-
5
633
FQ216406.1
-
5
635
FQ216495.1
-
5
632
FQ216618.1
-
5
632
FQ216655.1
-
5
633
FQ216733.1
-
5
634
FQ216774.1
-
5
632
FQ216813.1
-
5
631
FQ216851.1
-
5
632
FQ216913.1
-
5
633
FQ217406.1
-
5
632
FQ218561.1
-
5
632
FQ218832.1
-
5
632
FQ218943.1
-
5
632
FQ219025.1
-
5
635
FQ219184.1
-
5
636
FQ219389.1
-
5
632
FQ219457.1
-
5
632
FQ219678.1
-
5
632
FQ219815.1
-
5
632
FQ219817.1
-
5
632
FQ219837.1
-
5
632
FQ219855.1
-
5
632
FQ219923.1
-
5
632
FQ220008.1
-
5
632
FQ220051.1
-
5
632
FQ220077.1
-
5
632
FQ220115.1
-
5
632
FQ220181.1
-
5
632
FQ220201.1
-
5
630
FQ220213.1
-
5
632
FQ220247.1
-
5
633
FQ220282.1
-
5
634
FQ220284.1
-
5
632
FQ220302.1
-
5
632
FQ220338.1
-
5
632
FQ220345.1
-
5
633
FQ220370.1
-
5
632
FQ220406.1
-
5
632
FQ220413.1
-
5
632
FQ220420.1
-
5
633
FQ220444.1
-
5
632
FQ220445.1
-
5
628
FQ220449.1
-
5
632
FQ220514.1
-
5
632
FQ220527.1
-
5
632
FQ220533.1
-
5
664
FQ220542.1
-
5
632
FQ220545.1
-
5
632
FQ220634.1
-
5
632
FQ220654.1
-
5
632
FQ220663.1
-
5
632
FQ220684.1
-
5
633
FQ220769.1
-
5
632
FQ220787.1
-
5
632
FQ220801.1
-
5
632
FQ220833.1
-
5
632
FQ220838.1
-
5
632
FQ220888.1
-
5
632
FQ220932.1
-
5
633
FQ221146.1
-
5
632
FQ221380.1
-
5
632
FQ221556.1
-
5
630
FQ222171.1
-
5
632
FQ222181.1
-
5
632
FQ222327.1
-
5
632
FQ222772.1
-
5
632
FQ222906.1
-
5
632
FQ223121.1
-
5
632
FQ223487.1
-
5
632
FQ224547.1
-
5
632
FQ224834.1
-
5
632
FQ224846.1
-
5
632
FQ225049.1
-
5
632
FQ225170.1
-
5
632
FQ225608.1
-
5
632
FQ225654.1
-
5
634
FQ225860.1
-
5
634
FQ225886.1
-
5
632
FQ226306.1
-
5
634
FQ227381.1
-
5
632
FQ227522.1
-
5
627
FQ227719.1
-
5
632
FQ227837.1
-
5
632
FQ227937.1
-
5
633
FQ227941.1
-
5
632
FQ227942.1
-
5
752
FQ228548.1
-
5
632
FQ228995.1
-
5
632
FQ228996.1
-
5
632
FQ229139.1
-
5
634
FQ229159.1
-
5
632
FQ229164.1
-
5
633
FQ229208.1
-
5
632
FQ229241.1
-
5
632
FQ229244.1
-
5
632
FQ229248.1
-
3
512
FQ229272.1
-
5
632
FQ229273.1
-
5
633
FQ229303.1
-
5
632
FQ229337.1
-
5
632
FQ229352.1
-
5
632
FQ229365.1
-
5
632
FQ229397.1
-
5
632
FQ229408.1
-
5
633
FQ229433.1
-
5
632
FQ229448.1
-
5
632
FQ229521.1
-
5
632
FQ229547.1
-
5
632
FQ229554.1
-
5
632
FQ229566.1
-
5
628
FQ229610.1
-
5
632
FQ229626.1
-
5
629
FQ229726.1
-
5
632
FQ229740.1
-
5
632
FQ229742.1
-
5
632
FQ229776.1
-
5
632
FQ229808.1
-
5
632
FQ229815.1
-
5
632
FQ229822.1
-
5
632
FQ229830.1
-
5
634
FQ229883.1
-
5
634
FQ229929.1
-
5
632
FQ229932.1
-
3
530
FQ229952.1
-
5
632
FQ229974.1
-
5
632
FQ230005.1
-
5
632
FQ230055.1
-
5
631
FQ230127.1
-
5
632
FQ230267.1
-
5
635
FQ230943.1
-
5
632
FQ231445.1
-
5
634
FQ231525.1
-
5
632
FQ231591.1
-
5
634
FQ231710.1
-
5
633
FQ231733.1
-
5
633
FQ231897.1
-
5
634
FQ231967.1
-
5
632
FQ232029.1
-
5
633
FQ232154.1
-
5
632
FQ232165.1
-
5
632
FQ232310.1
-
5
631
FQ232319.1
-
5
633
FQ232697.1
-
5
634
FQ232767.1
-
5
632
FQ232951.1
-
5
632
FQ233152.1
-
5
631
FQ233377.1
-
5
632
FQ233473.1
-
5
632
FQ233732.1
-
5
632
FQ233737.1
-
5
632
FQ233808.1
-
5
632
FQ234195.1
-
5
632
FQ234341.1
-
5
632
FQ234613.1
-
5
632
FQ234733.1
-
5
632
FQ234735.1
-
5
522
FQ234863.1
-
5
632
FQ235063.1
-
5
632
FQ235079.1
-
5
632
FQ235259.1
-
5
632
X15096.1
-
5
632
Z29530.1
-
4
588
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_007475, BC030189, AK029816, BC011106, AK167240, and others.
Panther Classification:
Molecular Function -
ribosomal protein
translational protein
Gene Ontology Categories:
Function(s)
Process(es)
translational elongation
ribosome biogenesis
cellular response to interleukin-4
structural constituent of ribosome
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-7745c474b-dptlh:80/100.66.78.226:80.
git-commit: c3c9db9cc82507548abdd0ae6a4ade3f81f66292
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.20.0-2024.11.45-1.0