Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlst8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022404.2 | NP_071799.2 | 4 | 569 | ||
GenBank mRNA | AF051155.2 | - | 4 | 568 | ||
BC097319.1 | - | 4 | 247 | 7455095 |