Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022510.1 | NP_071955.1 | 10 | 1255 | ||
GenBank mRNA | BC063811.1 | - | 10 | 1271 | 6919340 | |
BC081801.1 | - | 10 | 1226 | 7192390 | ||
FQ209448.1 | - | 9 | 1110 | |||
FQ209532.1 | - | 9 | 1109 | |||
FQ209571.1 | - | 9 | 1109 | |||
FQ212557.1 | - | 9 | 1071 | |||
FQ212566.1 | - | 9 | 1111 | |||
FQ212656.1 | - | 9 | 1110 | |||
FQ212694.1 | - | 1227 | ||||
FQ212896.1 | - | 9 | 1108 | |||
FQ212979.1 | - | 7 | 901 | |||
FQ213045.1 | - | 9 | 1110 | |||
FQ213454.1 | - | 10 | 1263 | |||
FQ215017.1 | - | 9 | 1065 | |||
FQ215042.1 | - | 9 | 1111 | |||
FQ215264.1 | - | 9 | 1109 | |||
FQ215283.1 | - | 9 | 1109 | |||
FQ215533.1 | - | 9 | 1109 | |||
FQ215586.1 | - | 9 | 1110 | |||
FQ215649.1 | - | 9 | 1110 | |||
FQ216342.1 | - | 9 | 1110 | |||
FQ216573.1 | - | 9 | 1110 | |||
FQ216754.1 | - | 9 | 1109 | |||
FQ216792.1 | - | 9 | 1110 | |||
FQ218195.1 | - | 9 | 1110 | |||
FQ219327.1 | - | 9 | 1109 | |||
FQ219561.1 | - | 9 | 1111 | |||
FQ219701.1 | - | 9 | 1109 | |||
FQ219716.1 | - | 9 | 1110 | |||
FQ220231.1 | - | 9 | 1107 | |||
FQ220328.1 | - | 7 | 931 | |||
FQ221674.1 | - | 9 | 941 | |||
FQ226216.1 | - | 9 | 1110 | |||
FQ228709.1 | - | 9 | 1109 | |||
FQ228734.1 | - | 10 | 1260 | |||
FQ228797.1 | - | 9 | 1110 | |||
FQ228847.1 | - | 9 | 1109 | |||
FQ228910.1 | - | 9 | 1110 | |||
FQ228929.1 | - | 9 | 1110 | |||
FQ228930.1 | - | 9 | 1109 | |||
FQ229086.1 | - | 9 | 1110 | |||
FQ229087.1 | - | 9 | 1109 | |||
FQ229116.1 | - | 9 | 1110 | |||
FQ229475.1 | - | 6 | 742 | |||
FQ229723.1 | - | 7 | 849 | |||
FQ229849.1 | - | 9 | 1068 | |||
FQ230368.1 | - | 9 | 1108 | |||
FQ231653.1 | - | 9 | 1109 | |||
FQ232105.1 | - | 9 | 1109 | |||
FQ232562.1 | - | 9 | 1112 | |||
FQ234293.1 | - | 9 | 1109 | |||
FQ234521.1 | - | 9 | 1113 | |||
FQ234619.1 | - | 9 | 1110 | |||
X82180.1 | - | 10 | 1255 |