Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022511.2 | NP_071956.2 | 2 | 379 | ||
GenBank mRNA | BC062405.1 | - | 2 | 379 | 5623232 | |
FQ217736.1 | - | 2 | 413 | |||
FQ221987.1 | - | 2 | 413 | |||
FQ227505.1 | - | 2 | 414 | |||
FQ228171.1 | - | 2 | 413 | |||
FQ232047.1 | - | 2 | 412 | |||
FQ232161.1 | - | 2 | 369 | |||
FQ232713.1 | - | 2 | 383 | |||
FQ232749.1 | - | 2 | 378 | |||
FQ235292.1 | - | 2 | 414 | |||
X96967.1 | - | 2 | 262 |